NANOCOLOR® Photometric Tube Tests, AOX
Supplier: MACHEREY-NAGEL
Certificates
About this item
Photometric test kit for the determination of AOX in water samples.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers Precisely pre-dosed reagents with constant high quality from batch to batch Coloured pictogram instructions and easy handling of the tubes 16 mm (OD) tubes for easy pipetting
The test kit is evaluated in 16 mm (OD) tubes. It is preprogrammend on NANOCOLOR® photometers. Programming data for other spectrophotometers are available.
Sea water analysis: Yes Hazardous material: Yes Remark: Measuring range on NANOCOLOR VIS II
Delivery: Supplied in rugged box with 20 test tubes.
Specifications
- Model:MANA985007
- Platform:NANOCOLOR® tube tests
- Storage temperature:15…25 °C
- Shelf life (months):12
- Weight:2067 g including packaging