


Ammonium cerium(IV) nitrate ≥99.0%, AnalaR NORMAPUR® ACS analytical reagent
Catalog # VWRC86167.180
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Assay (oxidimetric):≥ 99.0 %
- Appearance Reddish-orange crystalline powder:Passes test
- Heavy metals (as Pb) :≤ 20 ppm
- Insoluble in dilute H₂SO₄ :≤ 0.05 %
- Cl (Chloride):≤ 100 ppm
- PO₄ (Phosphate) :≤ 0.02 %
- Fe (Iron) :≤ 50 ppm
- Na (Sodium) :≤ 50 ppm
Specification Test Results
About this item
:
:548.22 g/mol
Storage Temperature:Ambient
UN:1477
ADR:5.1,III
:GHS03
:GHS07
:GHS05
CAS Number:16774-21-3
Formula:(NH₄)₂Ce(NO₃)₆
MDL Number:MFCD00151121