Order Entry
Singapore
ContactUsLinkComponent
Anti-Coronavirus (SARS-CoV-2) (B) spike S2 Mouse Monoclonal Antibody
Anti-Coronavirus (SARS-CoV-2) (B) spike S2 Mouse Monoclonal Antibody
Supplier:  MP Biomedicals
Certificates
undefined
Anti-Coronavirus (SARS-CoV-2) (B) spike S2 Mouse Monoclonal Antibody
Supplier:  MP Biomedicals

Specifications

  • Catalog No:
  • ICNA08720412
  • ICNA08720411
  • Antigen name:
  • Coronavirus (SARS-CoV-2) (B) spike S2
  • Coronavirus (SARS-CoV-2) (B) spike S2
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1

Specifications