Disinfectant, Quaternary, TexQ™, Texwipe®
Supplier: Texwipe
Certificates
About this item
TexQ™ Disinfectant is an EPA-registered quaternary ammonium compound (quat) which serves as a one-step cleaner/disinfectant (effective against a broad spectrum of bacteria, virus and fungi, and inhibits the growth of mould and mildew and their odours) when used as directed.
TexQ™ Disinfectant is available in two solutions: Ready-to-Use trigger spray(TX650) and Concentrate gallon bottle (TX651).
Meets FDA guideline for medical device bacterial endotoxin limit of not more than 20 EU/device.
Both formulations have 25% more kill claims.
Ordering information: Each bottle includes a ready-to-use trigger sprayer and a set of 5 colour indicator dots for easy identification.