Wipes for general applications, WYPALL* colour coded
Colour coded wipers (regular duty) suitable for a wide range of cleaning tasks. Ideal for use in the food service general cleaning and kitchen preparation and cleaning.
- Cloth-like comfort for more effective clean up
- Excellent speed of absorbency, absorbing up to five times their own weight
- Reusable and easily rinsed clean
Colour coding for task segregation, reduces risks of cross—contamination, available in four colours.
Blue for general cleaning, green for kitchen and food preparation, red for toilets and washrooms and yellow for infectious and isolation areas.
: Endorsed by HACCP International (Food zone secondary).