Order Entry
Singapore
ContactUsLinkComponent
Wipes for WETTASK™ system
Wipes for WETTASK™ system
Catalog # KIMB06001
Supplier:  KIMBERLY CLARK
Wipes for WETTASK™ system
Catalog # KIMB06001
Supplier:  KIMBERLY CLARK

Specifications

  • Packed:
    6 rolls/case
  • Description:
    Small bucket with case
  • L×W:
    318×305 mm
  • Pk:
    6 Roll

Specifications

About this item

The Kimtech™ Prep Wipers for the WetTask System, for Solvents, is built for critical tasks, such as preparing cleaning surfaces.

  • Cuts solvent usage by 20% using the Wettask closed bucket
  • Conveniently portable
  • Eliminate the need for laundering contaminated cloth rags

The enclosed system also helps eliminate exposure to chemical vapors and splashes. The Wettask closed bucket and wet wiping system can rreduce VOC emissions for a safer, more cost effective workplace.

Packaging: 6 rolls / case, 95 sheets / roll, 570 sheets / case.