
Chloral hydrate 98.5-101% (by acidimetry), GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:302-17-0
:
Formula:Cl₃CCH(OH)₂
MDL Number:MFCD00044479
:165.4 g/mol
Melting Pt:52 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Assay (acidimetric):98,5 – 101 %
- Identification:Passes test
- Acidity:Passes test
- Appearance of solution (10%, H₂O):Clear and colourless
- pH (10%, H₂O):3,5 – 5,5
- Chlorides (Cl):Max. 0,01 %
- Heavy metals (as Pb):Max. 0,002 %
- Toluene:Max. 0,089 %
- Chloral alcoholate:Passes test
- Substances darkened by H₂SO₄:Passes test
- Residue on ignition:Max. 0,1 %