HPLC columns, HALO® ES-CN, BIOCLASS
Supplier: ADVANCED MATERIALS TECHNOLOGY
Certificates
About this item
HALO® BIOCLASS ES-CN Peptide columns made with innovative Fused-Core® particle technology are ideal for both ultrafast and ultrahigh resolution separations of peptides and polypeptides up to 20 kDa. HALO® peptide columns offer comparable peak capacity to sub-2 μm non-core columns at 40 to 50% back pressure (2,7 μm). HALO® 160 Å peptide columns are optimised for high performance in HPLC, UHPLC, and LCMS peptide applications.
- Ideal for tryptic digests, post-translational modifications, (PTM)s; polypeptides
- HALO® Peptide ES-CN offers improved retention for polar peptides
- HALO® Peptide columns can be used in series to increase peak capacity for UHPLC and HPLC analyses of complex tryptic digest samples
- Particle Sizes: 2,7 or 5 μm superficially porous particle silica ('solid-core', 'core-shell')
Alternate selectivities with HALO® Peptide ES-C18, ES-CN, and Phenyl-Hexyl phases.
HALO® guard columns are available for this phase.
Specifications
- Catalog No:
- AMTE95124-904
- AMTE92124-504
- AMTE95120-404
- AMTE95123-904
- AMTE95123-204
- AMTE92120-604
- AMTE92124-404
- AMTE95122-704
- AMTE92120-704
- AMTE95123-104
- AMTE92120-404
- AMTE95122-904
- AMTE91226-704
- AMTE92124-204
- AMTE92124-304
- AMTE95122-604
- AMTE95120-904
- AMTE92123-904
- AMTE91226-404
- AMTE92124-104
- AMTE91226-604
- AMTE92121-904
- AMTE95123-704
- AMTE95123-504
- AMTE92121-604
- AMTE92121-704
- AMTE95123-604
- AMTE92124-904
- AMTE95123-304
- AMTE95120-704
- AMTE92121-404
- AMTE95123-404
- AMTE92124-604
- AMTE92124-704
- AMTE95120-604
- AMTE95124-504
- AMTE92122-404
- AMTE95124-304
- AMTE92123-104
- AMTE92122-504
- AMTE95124-404
- AMTE92123-204
- AMTE92122-204
- AMTE95124-104
- AMTE92122-704
- AMTE92122-304
- AMTE92122-904
- AMTE95124-204
- AMTE92122-104
- AMTE92122-604
- AMTE95122-404
- AMTE95122-504
- AMTE91225-704
- AMTE95122-204
- AMTE92123-704
- AMTE95122-304
- AMTE91225-404
- AMTE92123-504
- AMTE95124-704
- AMTE92123-604
- AMTE95122-104
- AMTE91225-604
- AMTE92123-304
- AMTE95124-604
- AMTE92123-404
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- Temperature limit:
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- 90 °C (pH 1); 40 °C (pH 8)
- USP code:
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10