HPLC columns, HALO® Penta-HILIC
HALO® Penta-HILIC columns made with innovative Fused-Core® particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO® 90 Å Penta-HILIC columns are optimised for high performance in HPLC, UHPLC, and LCMS small molecule applications.
- Ideal for polar compounds poorly retained in reversed-phase modes
- Alternate selectivity for HILIC mode
- Excellent peak shape for basic compounds in HILIC mode
- Particle sizes: 2, 2,7 or 5 μm superficially porous particle silica ('solid-core', 'core-shell')
HALO® guard columns are available for this phase.
- Catalog No:
- AMTE98216-405
- AMTE91812-905
- AMTE95813-705
- AMTE91813-405
- AMTE95812-705
- AMTE95814-305
- AMTE91812-405
- AMTE91813-905
- AMTE92812-105
- AMTE95812-305
- AMTE92813-105
- AMTE95813-305
- AMTE95814-705
- AMTE92814-505
- AMTE92812-505
- AMTE92813-505
- AMTE92811-405
- AMTE92814-105
- AMTE98215-405
- AMTE98216-605
- AMTE95813-905
- AMTE95814-405
- AMTE91812-105
- AMTE91813-105
- AMTE95812-905
- AMTE92814-605
- AMTE95814-905
- AMTE95810-605
- AMTE91812-505
- AMTE91813-505
- AMTE92813-205
- AMTE95813-405
- AMTE92810-605
- AMTE92812-205
- AMTE95812-405
- AMTE92813-605
- AMTE92812-605
- AMTE92814-205
- AMTE98215-605
- AMTE95812-505
- AMTE95814-105
- AMTE95813-105
- AMTE95814-505
- AMTE92814-305
- AMTE91813-205
- AMTE92813-305
- AMTE95813-505
- AMTE92814-705
- AMTE91812-205
- AMTE92811-605
- AMTE91813-605
- AMTE95812-105
- AMTE92813-705
- AMTE91812-605
- AMTE95810-705
- AMTE92812-305
- AMTE92810-705
- AMTE92812-705
- AMTE98215-705
- AMTE91812-705
- AMTE95812-605
- AMTE91813-305
- AMTE95813-205
- AMTE95814-205
- AMTE92811-705
- AMTE91813-705
- AMTE95812-205
- AMTE92813-405
- AMTE95813-605
- AMTE92814-405
- AMTE95814-605
- AMTE95810-405
- AMTE91812-305
- AMTE98216-705
- AMTE92812-405
- AMTE92813-905
- AMTE92814-905
- AMTE92810-405
- AMTE95810-905
- AMTE92812-905
- Endcapped:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- pH range:
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- Temperature limit:
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- 60 ⁰C (pH 2); 40 ⁰C (pH 9)
- USP code:
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95
- L95