Order Entry
Singapore
ContactUsLinkComponent
VWR®, Variable Speed Rocker
VWR®, Variable Speed Rocker
Catalog # VWRU75832-308
Supplier:  VWR Collection
undefined
VWR®, Variable Speed Rocker
Catalog # VWRU75832-308
Supplier:  VWR Collection

Specifications

  • Description:
    2D variable speed rocker
  • Electrical:
    115 or 230 V; 50 - 60 Hz; 50 W
  • Load capacity:
    3,62 kg
  • Tilt angle:
    0 to 30° (Varaible)
  • Dimensions:
    355,6×304,8×203,2
  • Speed:
    2 - 32 min⁻¹ (Variable)
  • Weight:
    3,40 kg
  • Pk:
    1

Specifications

About this item

Designed to provide precise speed and tilt angles for a broad range of molecular and biological mixing applications.

  • Variable speed control up to 32 min⁻¹
  • Easily adjustable tilt angle, from 0 to 30°
  • Large 355,6×304,8 mm work surface incudes non-skid rubber pad
  • Suitable for use in the cold room

With variable speed control and adjustable tilt angles, the VWR® Rocker is ideal for many different applications including, gel staining, washing, antibody staining, hybridizations, more.

A large platform is included with a non-skid surface, ideal for securing all popular laboratory vessels. A powerful brushless motor drive provides a smooth and consistent rocking motion and speed is adjustable to provide rocking from gentle washing to aggressive mixing.

Certifications: CE Marked.
Ordering information: VWR® 2D Variable Speed Rocker comes with a two-year warranty.