HPLC columns, HALO® Diphenyl, BIOCLASS
Supplier: ADVANCED MATERIALS TECHNOLOGY
Certificates
About this item
HALO® BIOCLASS Diphenyl 400 Å protein columns made with innovative large pore Fused-Core® particle technology provide narrower peaks and better recoveries for high resolution separations of large biomolecules such as mAbs, versus smaller pore size columns. HALO® 400 Å columns are optimised for high performance in HPLC, UHPLC, and LCMS large molecule applications delivering excellent reproducibility and column lifetimes.
- Ideal for large protein analysis such as mAbs and polypeptides
- Well suited for high speed protein separations
- Exceptional recovery performance and temperature stability from 40 to 90 °C
- Outstanding low bleed performance
Alternate selectivities with HALO® Protein C4, Diphenyl and ES-C18 phases.
HALO® guard columns are available for this phase.
Specifications
- Catalog No:
- AMTE93412-626
- AMTE93412-126
- AMTE93414-426
- AMTE93414-626
- AMTE93412-726
- AMTE93412-426
- AMTE93414-726
- AMTE93414-126
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- Temperature limit:
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- 90 °C (pH 2); 40 °C (pH 9)
- USP code:
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11