HPLC columns, HALO® ES-C18, BIOCLASS
HALO® BIOCLASS ES-C18 400 Å Protein columns made with innovative large pore Fused-Core® particle technology provide narrower peaks and better recoveries for high resolution separations of large biomolecules such as mAbs, versus smaller pore size columns. HALO 400 Å columns are optimised for high performance in HPLC, UHPLC, and LCMS large molecule applications delivering excellent reproducibility and column lifetimes.
- Ideal for large protein analysis such as mAbs and polypeptides
- Well suited for high speed protein separations
- Outstanding low bleed performance and high temperature stability
Alternate selectivity’s with HALO® Protein C4, Diphenyl and ES-C18 phases.
HALO® guard columns are available for this phase.
- Catalog No:
- AMTE93410-702
- AMTE93413-402
- AMTE93412-202
- AMTE93412-302
- AMTE94316-702
- AMTE93413-202
- AMTE93412-102
- AMTE93414-402
- AMTE93414-502
- AMTE93413-302
- AMTE94316-402
- AMTE93412-902
- AMTE93414-202
- AMTE93413-102
- AMTE93411-702
- AMTE94316-602
- AMTE93414-302
- AMTE93410-602
- AMTE94315-602
- AMTE94315-702
- AMTE93410-402
- AMTE93412-702
- AMTE93414-102
- AMTE93412-502
- AMTE93414-902
- AMTE93413-702
- AMTE93413-902
- AMTE93412-602
- AMTE94315-402
- AMTE93412-402
- AMTE93414-702
- AMTE93411-602
- AMTE93413-502
- AMTE93413-602
- AMTE93411-402
- AMTE93414-602
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- Temperature limit:
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- 90 ⁰C (pH 1); 40 ⁰C (pH 8)
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1