HPLC columns, HALO® diphenyl, BIOCLASS
Supplier: ADVANCED MATERIALS TECHNOLOGY
Certificates
About this item
HALO® BIOCLASS Diphenyl 1000 Å protein columns made with innovative large pore Fused-Core® particle technology provide best in class performance for high resolution separations and better recoveries of large biomolecules such as mAbs, versus smaller pore size columns.
- Ideal for mAbs, ADC's, and and large proteins due to unrestricted access to the bonded phase
- Best in class resolution for full characterisation of mAbs and their variants
- Exceptional recovery performance and temperature stability from 40 to 90 °C
- Outstanding low bleed performance
HALO 1000 Å columns are optimized for high performance in HPLC, UHPLC, and LCMS large molecule applications delivering excellent reproducibility and column lifetimes.
Alternate selectivities with HALO® Protein C4, Diphenyl and ES-C18 phases.
HALO® guard columns are available for this phase.
Specifications
- Catalog No:
- AMTE92712-426
- AMTE92713-326
- AMTE92713-426
- AMTE92714-726
- AMTE92711-426
- AMTE92713-226
- AMTE92712-326
- AMTE92713-726
- AMTE97216-626
- AMTE92713-926
- AMTE92713-626
- AMTE92714-526
- AMTE92714-626
- AMTE97216-426
- AMTE92710-726
- AMTE92714-426
- AMTE97215-726
- AMTE92713-126
- AMTE92710-626
- AMTE92713-526
- AMTE92712-226
- AMTE92710-426
- AMTE92711-726
- AMTE97215-626
- AMTE92712-126
- AMTE92712-926
- AMTE92712-726
- AMTE92714-326
- AMTE97215-426
- AMTE92711-626
- AMTE92714-226
- AMTE92712-526
- AMTE92712-626
- AMTE97216-726
- AMTE92714-126
- AMTE92714-926
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- Temperature limit:
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- USP code:
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11
- L11