HPLC columns, HALO® C4, BIOCLASS
HALO® BIOCLASS C4 400 Å Protein columns made with innovative large pore Fused-Core® particle technology provide narrower peaks and better recoveries for high resolution separations of large biomolecules such as mAbs, versus smaller pore size columns. These columns are optimised for high performance in HPLC, UHPLC, and LCMS large molecule applications delivering excellent reproducibility and column lifetimes.
- Ideal for large protein analysis such as mAbs and polypeptides
- Well suited for high speed protein separations
- Outstanding low bleed performance and high temperature stability
Alternate selectivity's with HALO® Protein C4, Diphenyl and ES-C18 phases.
HALO® guard columns are available for this phase.
- Catalog No:
- AMTE93410-614
- AMTE94316-714
- AMTE93410-714
- AMTE93412-214
- AMTE93413-314
- AMTE93412-314
- AMTE93414-414
- AMTE94316-614
- AMTE93411-714
- AMTE93413-114
- AMTE93413-214
- AMTE93412-114
- AMTE93414-214
- AMTE94315-714
- AMTE93414-314
- AMTE94316-414
- AMTE93412-914
- AMTE93411-614
- AMTE94315-414
- AMTE93410-414
- AMTE94315-614
- AMTE93414-114
- AMTE93412-714
- AMTE93414-914
- AMTE93413-714
- AMTE93412-514
- AMTE93412-614
- AMTE93413-914
- AMTE93413-514
- AMTE93413-614
- AMTE93414-714
- AMTE93412-414
- AMTE93414-514
- AMTE93414-614
- AMTE93413-414
- AMTE93411-414
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- Temperature limit:
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- 90 ⁰C (pH 2); 40 ⁰C (pH 9)
- USP code:
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26
- L26