HPLC columns, HALO® HILIC
HALO® HILIC columns made with innovative Fused-Core® particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO® 90 Å HILIC columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
- Ideal for polar analytes
- Alternate mode to reversed-phase modes
- Can be used in HILIC and normal-phase modes
- Particle sizes: 2, 2,7 or 5 μm superficially porous particle silica ('solid-core', 'core-shell')
HALO® guard columns are available for this phase.
- Catalog No:
- AMTE92814-201
- AMTE98216-401
- AMTE91812-901
- AMTE95812-701
- AMTE95813-701
- AMTE91813-401
- AMTE92813-101
- AMTE95813-301
- AMTE95814-301
- AMTE95812-301
- AMTE92812-101
- AMTE91813-901
- AMTE92813-501
- AMTE92814-501
- AMTE95814-701
- AMTE91812-401
- AMTE92812-501
- AMTE92814-101
- AMTE92812-701
- AMTE98215-401
- AMTE98216-601
- AMTE91813-101
- AMTE95813-901
- AMTE91812-101
- AMTE91813-501
- AMTE95812-901
- AMTE95814-401
- AMTE91812-501
- AMTE95810-601
- AMTE92812-201
- AMTE95812-401
- AMTE95813-401
- AMTE92813-201
- AMTE92814-601
- AMTE95814-901
- AMTE92810-601
- AMTE92812-601
- AMTE92813-601
- AMTE92812-901
- AMTE98215-601
- AMTE95814-101
- AMTE95812-501
- AMTE92814-301
- AMTE95814-501
- AMTE91812-201
- AMTE91813-201
- AMTE95813-101
- AMTE92814-701
- AMTE95810-701
- AMTE91812-601
- AMTE92811-601
- AMTE91813-601
- AMTE95812-101
- AMTE92813-301
- AMTE95813-501
- AMTE92810-701
- AMTE92812-301
- AMTE92813-701
- AMTE92811-401
- AMTE98215-701
- AMTE95812-601
- AMTE95814-201
- AMTE91812-701
- AMTE95813-201
- AMTE95814-601
- AMTE92814-401
- AMTE91813-301
- AMTE95813-601
- AMTE92813-401
- AMTE92814-901
- AMTE91812-301
- AMTE95810-401
- AMTE95812-201
- AMTE92811-701
- AMTE91813-701
- AMTE92813-901
- AMTE98216-701
- AMTE95810-901
- AMTE92810-401
- AMTE92812-401
- pH range:
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- 1 - 8
- Temperature limit:
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- 60 °C (pH 1); 40 °C (pH 8)
- USP code:
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3
- L3