Hydrochloric acid 35,0 - 37,0%, Finyte®, J.T.Baker®
Catalog # BAKR9625-69
Supplier: Avantor
CAS Number:
Specifications
- Pk:186,4 L
- Seq:5495
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (as HCl) (by acid-base titrn):35.0 - 37.0 %
- Color (APHA):≤ 5
- Residue after Ignition:≤ 3 ppm
- Extractable Organic Substances:≤ 3 ppm
- Bromide (Br):≤ 0.005 %
- Free Halogen (as Cl₂):Passes Test
- Phosphate (PO₄):≤ 0.03 ppm
- Sulfate (SO₄):≤ 0.3 ppm
- Sulfite (SO₃):≤ 0.4 ppm
- Trace Impurities - Aluminum (Al):≤ 5.0 ppb
- Arsenic and Antimony (as As):≤ 3.0 ppb
- Trace Impurities - Barium (Ba):≤ 5.0 ppb
- Trace Impurities - Beryllium (Be):≤ 1.0 ppb
- Trace Impurities - Bismuth (Bi):≤ 5.0 ppb
- Trace Impurities - Boron (B):≤ 5.0 ppb
- Trace Impurities - Cadmium (Cd):≤ 5.0 ppb
- Trace Impurities - Calcium (Ca):≤ 5.0 ppb
- Trace Impurities - Chromium (Cr):≤ 5.0 ppb
- Trace Impurities - Cobalt (Co):≤ 5.0 ppb
- Trace Impurities - Copper (Cu):≤ 5.0 ppb
- Trace Impurities - Gallium (Ga):≤ 5.0 ppb
- Trace Impurities - Germanium (Ge):≤ 5.0 ppb
- Trace Impurities - Gold (Au):≤ 5.0 ppb
- Heavy Metals (as Pb):≤ 50.0 ppb
- Trace Impurities - Indium (In):≤ 5.0 ppb
- Trace Impurities - Iron (Fe):≤ 5.0 ppb
- Trace Impurities - Lead (Pb):≤ 5.0 ppb
- Trace Impurities - Lithium (Li):≤ 5.0 ppb
- Trace Impurities - Magnesium (Mg):≤ 5.0 ppb
- Trace Impurities - Manganese (Mn):≤ 5.0 ppb
- Trace Impurities - Molybdenum (Mo):≤ 5.0 ppb
- Trace Impurities - Nickel (Ni):≤ 5.0 ppb
- Trace Impurities - Niobium (Nb):≤ 1.0 ppb
- Trace Impurities - Platinum (Pt):≤ 5.0 ppb
- Trace Impurities - Potassium (K):≤ 5.0 ppb
- Trace Impurities - Silicon (Si):≤ 5.0 ppb
- Trace Impurities - Silver (Ag):≤ 5.0 ppb
- Trace Impurities - Sodium (Na):≤ 5.0 ppb
- Trace Impurities - Strontium (Sr):≤ 5.0 ppb
- Trace Impurities - Tantalum (Ta):≤ 5.0 ppb
- Trace Impurities - Thallium (Tl):≤ 5.0 ppb
- Trace Impurities - Tin (Sn):≤ 5.0 ppb
- Trace Impurities - Titanium (Ti):≤ 5.0 ppb
- Trace Impurities - Vanadium (V):≤ 1.0 ppb
- Trace Impurities - Zinc (Zn):≤ 5.0 ppb
- Trace Impurities - Zirconium (Zr):≤ 5.0 ppb
- Particle Count - 0.5 µm and greater:≤ 150 par/ml
- Particle Count - 1.0 µm and greater:≤ 25 par/ml
Specification Test Results
About this item
Synonyms: Hydrochloric acid solution, Muriatic acid
:
:36.46
Melting Pt:–30 °C
EINECS:231-595-7
UN:1789
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324
Boiling Pt:110 °C (1013 hPa)
Density:1.18 g/cm³ (20 °C)
ADR:8 II
:Management of Change (MOC) category = R