

2-Propanol ≥99.9%, Finyte® for microelectronic, J.T.Baker®
Catalog # BAKR5873-65
Supplier: Avantor
CAS Number:
Specifications
- Pk:1 gal US
- Seq:8080
Specifications
Specification Test Results
- Assay (CH₃CHOHCH₃):≥99.9 %
- Color (APHA):≤6
- Residue after Evaporation:≤3000.0 ppb
- Solubility in H₂O:Passes Test
- Water (H₂O)(by Karl Fischer titrn):≤0.05 %
- Acidity (µeq/g):≤0.2
- Alkalinity (µeq/g):≤0.1
- Chloride (Cl):≤60.0 ppb
- Nitrate (NO₃):≤0.1 ppm
- Sulfate (SO₄):≤0.1 ppm
- Phosphate (PO₄):≤75.0 ppb
- Trace Impurities - Aluminum (Al):≤10.0 ppb
- Arsenic and Antimony (as As):≤10.0 ppb
- Trace Impurities - Barium (Ba):≤10.0 ppb
- Trace Impurities - Beryllium (Be):≤10.0 ppb
- Trace Impurities - Bismuth (Bi):≤10.0 ppb
- Trace Impurities - Boron (B):≤3.0 ppb
- Trace Impurities - Cadmium (Cd):≤10.0 ppb
- Trace Impurities - Calcium (Ca):≤10.0 ppb
- Trace Impurities - Chromium (Cr):≤10.0 ppb
- Trace Impurities - Cobalt (Co):≤10.0 ppb
- Trace Impurities - Copper (Cu):≤10.0 ppb
- Trace Impurities - Gallium (Ga):≤10.0 ppb
- Trace Impurities - Germanium (Ge):≤10.0 ppb
- Trace Impurities - Gold (Au):≤5.0 ppb
- Trace Impurities - Iron (Fe):≤5.0 ppb
- Trace Impurities - Lead (Pb):≤10.0 ppb
- Trace Impurities - Lithium (Li):≤10.0 ppb
- Trace Impurities - Magnesium (Mg):≤10.0 ppb
- Trace Impurities - Manganese (Mn):≤10.0 ppb
- Trace Impurities - Molybdenum (Mo):≤10.0 ppb
- Trace Impurities - Nickel (Ni):≤10.0 ppb
- Trace Impurities - Niobium (Nb):≤10.0 ppb
- Trace Impurities - Potassium (K):≤10.0 ppb
- Trace Impurities - Silicon (Si):≤10.0 ppb
- Trace Impurities - Silver (Ag):≤5.0 ppb
- Trace Impurities - Sodium (Na):≤10.0 ppb
- Trace Impurities - Strontium (Sr):≤10.0 ppb
- Trace Impurities - Tantalum (Ta):≤10.0 ppb
- Trace Impurities - Thallium (Tl):≤10.0 ppb
- Trace Impurities - Tin (Sn):≤10.0 ppb
- Trace Impurities - Titanium (Ti):≤10.0 ppb
- Trace Impurities - Vanadium (V):≤10.0 ppb
- Trace Impurities - Zinc (Zn):≤10.0 ppb
- Trace Impurities - Zirconium (Zr):≤10.0 ppb
- Particle Count - 0.5 µm and greater:≤25 par/ml
- Particle Count - 1.0 µm and greater:≤8 par/ml
Specification Test Results
About this item
:
:60.1 g/mol
Merck Index:13,05228
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0,786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
:Management of Change (MOC) category = R
REACH:01-2119457558-25
:GHS02
:GHS07
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674