

Perchloric acid ≥70%, AR® ACS, Macron Fine Chemicals™
Catalog # MACR3999-46
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Glass bottle, clear, PVC-coated
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- ACS - Assay (HClO₄):69.0 - 72.0 %
- Appearance (clear, colorless solution):Passes Test
- ACS - Chloride (Cl):≤ 0.001 %
- ACS - Color (APHA):≤ 10
- ACS - Heavy Metals (by ICP-OES):≤ 1 ppm
- ACS - Iron (Fe):≤ 0.0001 %
- ACS - Nitrogen Compounds (as N):≤ 0.001 %
- ACS - Residue after Ignition:≤ 0.003 %
- ACS - Silicate and Phosphate (as SiO₂):≤ 0.0005 %
- ACS - Sulfate (SO₄):≤ 0.001 %
Specification Test Results
About this item
:
:100.46 g/mol
Boiling Pt:160…181 °C (1013 hPa)
Melting Pt:–112,2 °C
Density:1,53…1.768 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-512-4
UN:1873
ADR:5.1,I
:Management of Change (MOC) category = N
:GHS03
:GHS05
CAS Number:7601-90-3
Formula:HClO₄
MDL Number:MFCD00011325