Order Entry
Singapore
ContactUsLinkComponent
Dextrose kit, anhydrous USP, J.T.Baker®
Dextrose kit, anhydrous USP, J.T.Baker®
Catalog # BAKR1919-X1
Supplier:  Avantor
CAS Number:  
undefined
Dextrose kit, anhydrous USP, J.T.Baker®
Catalog # BAKR1919-X1
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Seq:
    0075

Specifications

Specification Test Results

  • Kit Components Meet Requirements:
    Passes Test

Specification Test Results

About this item

 :
Merck Index:14,04459
Melting Pt:146 °C
Density:1,54 g/cm³ (20 °C)
 :180.16 g/mol
Storage Temperature:Ambient
EINECS:200-075-1
CAS Number:50-99-7
 :492-62-6
Formula:C₆H₁₂O₆
MDL Number:MFCD00148912