Ferric pyrophosphate soluble, pearls, purified, Macron Fine Chemicals™
Catalog # MACR5528-25
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pk:150 lb
- Seq:7191
Specifications
Specification Test Results
- Assay (Fe):10.5 - 12.5 %
- Solubility:Passes Test
- Identification A:Passes Test
- Identification B:Passes Test
- Ammonium Salts:Passes Test
- Arsenic (As):≤0.0008 %
Specification Test Results
About this item
:
:745.2100 g/mol
CAS Number:10058-44-3
Formula:Fe₄O₂₁P₆
MDL Number:MFCD00016091