


Acetic acid glacial ≥99.7% (by GC, corrected for water content), BAKER ANALYZED® ACS aldehyde-free, J.T.Baker®
Catalog # BAKR9517-80
Supplier: Avantor
CAS Number:
Specifications
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay (by GC, corrected for water):≥99.7 %
- Color (APHA):≤10
- Dilution Test:Passes Test
- Residue after Evaporation:≤0.001 %
- Acetic Anhydride ((CH₃CO)₂O):≤0.01 %
- Chloride (Cl):≤1 ppm
- Sulfate (SO₄):≤1 ppm
- Trace Impurities - Heavy Metals:≤0.5 ppm
- Trace Impurities - Iron (Fe):≤0.2 ppm
- Substances Reducing Dichromate:Passes Test
- Substances Reducing Permanganate:Passes Test
- Titrable Base (meq/g):≤0.0004
- Acetaldehyde:≤0.005 %
- Sensitivity:Passes Test
- Trace Impurities - Arsenic and Antimony (as As):≤50.0 ppb
- Trace Impurities - Boron (B):≤100.0 ppb
- Trace Impurities - Chromium (Cr):≤200.0 ppb
- Trace Impurities - Cobalt (Co):≤100.0 ppb
- Trace Impurities - Copper (Cu):≤100.0 ppb
- Heavy Metals (as Pb):≤500.0 ppb
- Trace Impurities - Lead (Pb):≤300.0 ppb
- Trace Impurities - Manganese (Mn):≤200.0 ppb
- Trace Impurities - Nickel (Ni):≤100.0 ppb
- Trace Impurities - Potassium (K):≤300.0 ppb
- Trace Impurities - Tin (Sn):≤100 ppb
- Trace Impurities - Titanium (Ti):≤300.0 ppb
- Trace Impurities - Zinc (Zn):≤200.0 ppb
Specification Test Results
About this item
:
:60.05 g/mol
Boiling Pt:118 °C (1013 hPa)
Melting Pt:16,4 °C
Density:1,05 g/cm³ (20 °C)
Flash Pt:38,5 °C
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8,II
:Management of Change (MOC) category = N
REACH:01-2119475328-30
:GHS02
:GHS05
:GHS07
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152