The Portable centrifuge kit by Heathrow Scientific offers a convenient way to spin down and prepare samples outside of a conventional laboratory environment.
- Kit enables immediate on-site testing for faster results
- Includes five year warranty
The centrifuge kit enables the processing of samples before deterioration can take place and can be used in remote environments where a normal electrical supply is not an option.
Conveniently carry the centrifuge and its components securely in the DuraPorter® transport box that has a specially designed insert to seat the centrifuge, all its accessories and incorporates a 16-place micro tube rack as well. The DuraPorter is transparent, so any samples contained can be viewed and their integrity checked.
With its ergonomic design, the Heathrow Scientific Sprout Mini Centrifuge is loaded with features that bring increased efficiency to the lab and in the field. Tool-free snapspin rotors and quick-spin options make it fast and easy to use and prevents loss of rotor tools.
The lid was functionally designed to fit in the palm of the hand for easy repeat opening and quick tube extraction. A weighted rubberised base eliminates potential movement and dampens vibration and noise. The Sprout is ideal for microfilter cell separations and HPLC samples, with adapters to accommodate all major microtube and PCR tube styles (1,5/2,0, 0,5, 0,2 ml) for customised use.
The Sprout® Mini Centrifuge, comes with its own 12 V car adapter and a 12 VDC power adapter for when electrical outlets are in reach.
- Catalog No:
- HEAT120569
- HEAT120570
- HEAT120568
- Max. speed:
- 6000 min⁻¹
- Max. RCF:
- 2000 ×g
- Power supply:
- Suitable for worldwide voltages, 100 - 240 VAC, 50/60 Hz, 0,6 A, ~7,3 W
- W×D×H:
- 153×128×104 mm