Order Entry
Singapore
ContactUsLinkComponent
Diethylenetriaminepentaacetic acid ≥99%, GPR RECTAPUR®
Diethylenetriaminepentaacetic acid ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Diethylenetriaminepentaacetic acid ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (calculated on dried basis):
    Min. 99 %
  • IR Spectrum:
    Passes test
  • Melting point:
    219 - 221 °C
  • Ignition residue (SO₄):
    Max. 0.1 %
  • Water:
    Max. 0.5 %
  • Fe (Iron):
    Max. 10 ppm
  • Pb (Lead):
    Max. 10 ppm

Specification Test Results