Order Entry
Singapore
ContactUsLinkComponent
Clamp-on breaker lockout
Clamp-on breaker lockout
Catalog # BRDY65397
Supplier:  Brady
undefined
Clamp-on breaker lockout
Catalog # BRDY65397
Supplier:  Brady
Specifications
  • Type:
    Electrical risk
  • Colour:
    Red
  • Description:
    Clamp-on breaker lockout
  • W×D×H:
    40,3×55,2×82,5 mm
  • Pk:
    1
Specifications
About this item
Clamp-on breaker lockouts are versatile and easy to install using the proprietary thumbwheel design.
  • No screwdrivers needed
  • Extremely versatile
  • Detachable cleat (included) expands the range of applicable breakers
Equipping with the proper lockout tools and warning devices can save lives, reduce lost employee time, and cut insurance costs.