Order Entry
Singapore
ContactUsLinkComponent
Industrial gloves, oil repellent, HyFlex® 11-953
 :  Ansell
undefined
Industrial gloves, oil repellent, HyFlex® 11-953
 :  Ansell
  • Catalog No:
  • ANSE11-953/9
  • ANSE11-953/10
  • ANSE11-953/6
  • ANSE11-953/7
  • ANSE11-953/8
  • Model:
  • 11-953
  • 11-953
  • 11-953
  • 11-953
  • 11-953
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Polyamide
  • Polyamide
  • Polyamide
  • Polyamide
  • Polyamide
  • Finish:
  • Palm nitrile coated
  • Palm nitrile coated
  • Palm nitrile coated
  • Palm nitrile coated
  • Palm nitrile coated
  • Gauge:
  • 13
  • 13
  • 13
  • 13
  • 13
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (4121A)
  • EN 388:2016 (4121A)
  • EN 388:2016 (4121A)
  • EN 388:2016 (4121A)
  • EN 388:2016 (4121A)
  • Food contact approved:
  • +
  • +
  • +
  • +
  • +