Serum-free media, PowerStem iPS2
Catalog # PANBP04-77140K
Supplier: PAN BIOTECH
CAS Number:
Specifications
- Media Format:Liquid
- Pk:500 mL
- Description:PowerStem iPS2
- Shelf life:12 Months
Specifications
About this item
PowerStem iPS2 is a specialised serum-free, fully defined medium for cultivation and expansion of human induced pluripotent stem cells (iPS cells). Human induced pluripotent stem cells have the capacity to differentiate into all of the somatic cell types and therefore hold great promise for regenerative medicine. Even after long-term culture (>50 passages) iPS cells retain a normal karyotype and a stable proliferating rate.
- Suitable for the cultivation of human induced pluripotent stem cells (iPS cells), while maintaining an undifferentiated state
- Store at –20 °C
It contains purified and recombinant proteins, lipids, salts, amino acids, trace elements, attachment factors, hormones and growth factors in an optimised formulation. It is fully defined and contains no animal-derived components.