Serum-free media, PowerStem iPS1
PANBP04-77130K
:
- Media Format:Liquid
- Pk:500 mL
- Description:PowerStem iPS1
- Shelf life:12 Months
PowerStem iPS1 is a fully defined, specialised serum-free medium for the cultivation and expansion of human induced pluripotent stem cells (iPS cells). Induced pluripotent stem cells have the capacity to differentiate into all of the somatic cell types and therefore hold great promise for regenerative medicine. Even after long-term culture (>50 passages) iPS cells retain a normal karyotype and a stable proliferating rate.
- Suitable for the cultivation of human induced pluripotent stem cells (iPS cells),while maintaining an undifferentiated state
- Store at –20 °C
It contains purified proteins, lipids, salts, amino acids, trace elements, attachment factors, hormones and growth factors in an optimised formulation. It is free of animal-derived components.