Order Entry
Singapore
ContactUsLinkComponent
Cell culture supplements, Panexin BMM serum substitute
Cell culture supplements, Panexin BMM serum substitute
Catalog # PANBP04-951SA2
Supplier:  PAN BIOTECH
CAS Number:  
Cell culture supplements, Panexin BMM serum substitute
Catalog # PANBP04-951SA2
Supplier:  PAN BIOTECH
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Description:
    Panexin BMM serum substitute

Specifications

About this item

Panexin BMM is a fully defined serum substitute for the cultivation of macrophages from mouse bone marrow (murine bone marrow derived macrophages, BMM) under serum- free conditions.

  • Ready-to-use
  • Store at –20 °C

It contains purified proteins, lipids, salts, amino acids, trace elements, attachment factors and hormones in an optimised formulation. It contains no growth factors, undefined hydrolysates or lysates (e. g. Peptones).