Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cell Culture Supplements and Additives
Tiamulin
Tiamulin
Tiamulin
Catalog # PANBP06-12025
Supplier:
PAN BIOTECH
CAS Number:
Expand image
Tiamulin
Catalog # PANBP06-12025
Supplier:
PAN BIOTECH
CAS Number:
Specifications
Pk:
25
Description:
Tiamulin, 5 ml
Storage temperature:
–20 °C
Specifications
About this item
Product Details
Concentration: 1 mg/ml
Sterile filtered
Store at ‒20 °C
Product Details