Dimethyl sulphoxide ≥99.5%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
Catalog # BJAA081-4
Supplier: BURDICK JACKSON
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
Specifications
Specification Test Results
- GC Analysis:99.5 % min.
- Refractive Index (20ºC):1.4770 - 1.4800
- UV Absorbance @ 275 nm:0.5 AU max.
- UV Absorbance @ 300 nm:0.2 AU max.
- UV Absorbance @ 350 nm:0.02 AU max.
- UV Absorbance @ 400 nm:0.005 AU max.
- UV Cutoff:268 nm max.
- Water by Karl Fischer Titration:0.04 % max.
Specification Test Results
About this item
:
:78.14 g/mol
Merck Index:13,03285
Boiling Pt:189 °C
Melting Pt:18,5 °C
Density:1,104 g/cm³ (20 °C)
Flash Pt:28,5 %(V)
Storage Temperature:Ambient
EINECS:200-664-3
CAS Number:67-68-5
Formula:(CH₃)₂SO
MDL Number:MFCD00002089