Lead(II) acetate basic ACS
Glass bottle for solids, 2 kg
Supplier: Thermo Fisher Scientific
Lead(II) acetate basic ACS
Catalog # ALFA36393.A3
Supplier: Thermo Fisher Scientific
CAS Number: 1335-32-6
Jump to:
Product Details & Documents
CAS No.: 1335-32-6 Molecular Formula: C4H6O4Pb.2Pb(OH)2 Formula Weight: 807.71 Physical Form: Powder Sensitivity: Air Sensitive Merck Reference: 14,5419 MDL No.: MFCD00013041
Synonyms: Horne′s compound, Lead(II) hydroxide acetate, Acetic acid lead (II) salt basic, Lead subacetate
- CAS Number:1335-32-6
- Formula:(CH₃COO)₂Pb·2Pb(OH)₂
- MDL Number:MFCD00011154
- Molecular Weight:807.72 g/mol
- Merck Index:14,5419
- Storage Temperature:Ambient
- EINECS:215-630-3
- UN:1616
- ADR:6.1,III
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:2 kg



