Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
ω-Bromo-2-nitroacetophenone
ω-Bromo-2-nitroacetophenone ≥98%
ω-Bromo-2-nitroacetophenone ≥98%
ω-Bromo-2-nitroacetophenone ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
ω-Bromo-2-nitroacetophenone ≥98%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
6851-99-6
:
Formula:
C₈H₆BrNO₃
MDL Number:
MFCD00010294
:
244.04
Melting Pt:
53…56 °C
Storage Temperature:
Ambient
UN:
3261
ADR:
8,II
:
GHS05
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH30275
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation