Order Entry
Singapore
ContactUsLinkComponent
Cell dissociation solution, EDTA
Cell dissociation solution, EDTA
  PANBP10-15100
 :  
Cell dissociation solution, EDTA
  PANBP10-15100
 :  

 

  • Pk:
    100 mL
  • Description:
    Cell dissociation solution, EDTA
  • Storage conditions:
    2...8 °C
  • Formulation:
    EDTA 1:5000 (1X), 0,2 g/L in PBS, without calcium and magnesium

 

 

This sterile solution has 24 months shelf life. The shelf life commences with date of production.

  • Dissociation of tissue and cell monolayer cultures