
Dodecyl-3,4,5-trihydroxybenzoate
Catalog # EDQMY0000365
Supplier: EDQM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:30 mg
- Seq:2345
Specifications
About this item
:
:338.44 g/mol
Melting Pt:94…97 °C
EINECS:214-620-6
:GHS07
CAS Number:1166-52-5
Formula:C₁₉H₃₀O₅
MDL Number:MFCD00002195