Cell culture media, IPL-41 insect medium
Catalog # PANBP03-9205S1
Supplier: PAN BIOTECH
CAS Number:
Specifications
- Pk:5 L
- Description:IPL-41 insect medium
Specifications
About this item
IPL-41 is primarly used for the growth and maintenance of lepidopteran cells and for the propagation of viruses in these cells lines. The medium is also used for long time culture of baculo-virus infected spodoptera cells.
- Sterile medium
- Store at ‒20 °C
This medium contains L-Glutamine and 0,35 g/L NaHCO₃ powder.