Cell culture supplements, Panexin, serum replacement
Catalog # PANBP04-95100
Supplier: PAN BIOTECH
CAS Number:
Specifications
- Pk:100 mL
- Description:Panexin, serum replacement
Specifications
About this item
Panexin is a chemically defined serum substitute for the cultivation of adherent and non-adherent cells under serum-free conditions.
- Sterile solution ready for use
- End concentration of 2 to 8%
- Supports growth of many cell types
- Sterile solution
- Store at ‒20 °C
Panexin contains cleaned protein, lipids, salts, amino acids, trace elements, attachment factors and hormones in an optimised recipe. It does not contain any undefined hydrolysates or lysates.