L-proline
Catalog # PANBP60-17050100
Supplier: PAN BIOTECH
CAS Number:
Specifications
- Pk:100 g
- Description:L-proline, powder
Specifications
About this item
This is suitable for cell biology.
- Sterile
- Stored at 2 to 8 °C
Amino acids are crucial for cell culture. Cells cannot synthesise essential amino acids and have to be added to cell culture, while non-essential-amino acids relieve the intracellular metabolism and help cells grow better under the same circumstances.