MEM vitamin supplement 100X
Catalog # PANBP08-41001P
Supplier: PAN BIOTECH
CAS Number:
Specifications
- Pk:1 L
- Description:MEM vitamin supplement (100X), powder
Specifications
About this item
The availability of vitamins is extremely important for the cells, since the cells usually cannot synthesise the vitamins.
- Stored at 2 to 8 °C
Vitamins act as co-factors for many enzymes and are essential for their function. The absence of vitamins in culture may lead to decrease in cell growth, cell death or loss of productivity.