Poly-L-lysine solution
PANBP08-48350
:
- Pk:50 mL
- Description:Poly-L-lysine solution, 0,01%, BioReagent
- Molecular weight:150000 - 300000
This is suitable for cell culture.
- Sterile
- Stored at 2 to 8 °C
Amino acids are crucial for cell culture. Cells cannot synthesise essential amino acids and have to be added to cell culture, while non-essential-amino acids relieve the intracellular metabolism and help cells grow better under the same circumstances.