Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-6-nitrophenol
2-Bromo-6-nitrophenol ≥97%
2-Bromo-6-nitrophenol ≥97%
2-Bromo-6-nitrophenol ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
2-Bromo-6-nitrophenol ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
13073-25-1
:
Formula:
C₆H₄BrNO₃
MDL Number:
MFCD02683216
:
218.01 g/mol
Melting Pt:
66…69 °C
:
GHS06
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAH27326
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation