γ-(N-Morpholino)propanesulphonic acid (MOPS) ≥99% (dry basis), Ultrapure Bioreagent for molecular biology, J.T.Baker®
Catalog # BAKR4004-01
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Poly bottle, white, blue T/E cap
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99 %
- Assay (C₇H₁₅NO₄S) (dried basis):≥99 %
- DNase Activity (None Detected):Passes Test
- RNase Activity:Passes Test
- Protease Activity:Passes Test
- Appearance (white to slightly pink powder to crystalline powder):Passes Test
- pH of 1% Solution at 25°C:2.5 - 4.5
- Solution (10%) in Water:Passes Test
- Loss on Drying:≤1 %
- Absorbance of a 0.1M Solution (1-cm path vs water) at - 260 nm:≤0.020
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
Specification Test Results
About this item
Synonyms: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
:
:209.27 g/mol
EINECS:214-478-5
:GHS07
Storage Temperature:Ambient
CAS Number:1132-61-2
Formula:C₇H₁₅NO₄S
MDL Number:MFCD00006183
Melting Pt:> 250 °C
Flash Pt:110 °C (closed cup)
Merck Index:13,06288