

N-Methyl-2-pyrrolidone (NMP) ≥99.5%, BioSyn™ for biosynthesis, for applications requiring low water content, Burdick & Jackson™
BJAABB305-4
:
- Pack type:Glass bottle
- Pk:16 L
- Seq:5254
- Amines (as dimethylamine):3 ppm max.
- Water by Karl Fischer Titration:200 ppm max.
- GC Analysis:99.5 % min.
- Refractive Index (20ºC):1.4680 - 1.4720
- Residue:5 mg/L max.
- UV Absorbance @ 300 nm:0.5 AU max.
- UV Absorbance @ 325 nm:0.1 AU max.
- UV Absorbance @ 350 nm:0.03 AU max.
- UV Absorbance @ 400 nm:0.01 AU max.
- UV Cutoff:285 nm max.
:
:99.13 g/mol
:13,06140
:202 °C (1013 hPa)
:-24 °C
:1,033 g/cm³ (20 °C)
:91 °C
:212-828-1
:GHS07
:GHS08
:Ambient
:872-50-4
:C₅H₉NO
:MFCD00003193