Isopropyl myristate, B&J Brand™ for sterility testing of ophthalmic ointments, Burdick & Jackson™
Catalog # BJAA225-4
Supplier: BURDICK JACKSON
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:4 L
- Seq:5162
Specifications
Specification Test Results
- Appearance:Passes test
- Identification by IR:Passes test
- pH of Water Extract:6.5 min.
Specification Test Results
About this item
:
:270.46 g/mol
Merck Index:13,05235
Boiling Pt:193 °C (26,7 hPa)
Melting Pt:–3 °C
Density:0,853 g/cm³ (20 °C)
Flash Pt:164 °C
Storage Temperature:Ambient
EINECS:203-751-4
CAS Number:110-27-0
Formula:C₁₇H₃₄O₂
MDL Number:MFCD00008982