Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Buffers
Electrophoresis Buffers
Running Buffers
Tris-acetate-EDTA (TAE) buffer
Tris-acetate-EDTA (TAE) buffer
Tris-acetate-EDTA (TAE) buffer
Catalog # PANBP60-16911000
Supplier:
PAN BIOTECH
CAS Number:
Expand image
Tris-acetate-EDTA (TAE) buffer
Catalog # PANBP60-16911000
Supplier:
PAN BIOTECH
CAS Number:
Specifications
Pk:
1.000 mL
Description:
TAE buffer, 50X
Specifications
About this item
Product Details
Product Details