Protein-free media, Panserin H8000
Catalog # PANBP04-718000
Supplier: PAN BIOTECH
CAS Number:
Specifications
- Media Format:Liquid
- Pk:500 mL
- Description:Panserin H8000, with L-Glutamine and cholesterol
- Seq:02
Specifications
About this item
Panserin H8000 is a protein-free, ready-to-use medium for an optimised growth of cholesterol-dependent myeloma and hybridoma cell lines in suspension culture for the production of monoclonal antibodies.
- Suitable for spinner cultures, roller bottles and bioreactors
- Store at 2 to 8 °C
It consists of a balanced mixture of salts, amino acids, vitamins, trace elements, hormones, bioavailable cholesterol and is enriched with selected herbal hydrolysates.