Serum-free media, Panserin C6000
PANBP04-716000
:
- Media Format:Liquid
- Pk:500 mL
- Description:Panserin C6000, with L-Glutamine
Panserin C6000 is a protein-free ready to use medium for an optimized growth of CHO-cells (Chinese Hamster Ovary) and their recombinant derivates in suspension culture. These cells are often used for the production of recombinant proteins for diagnostic or therapeutic purposes.
- Suitable for spinner cultures, roller bottles and tissue culture flasks and bioreactors
- Store at 2 to 8 °C
It consists of a balanced mixture of salts, amino acids, vitamins, trace elements, hormones and is enriched with select herbal hydrolysates for an optimised growth of CHO-cells in suspension culture. As Panserin C6000 is free of animal or human components it is predestined for the use in sensitive production areas (e.g. production of diagnostic or therapeutic tools) where safety requirements prohibit the use human or animal components.