Serum-free media, Panserin PX40
PANBP04-710PX40
:
- Media Format:Liquid
- Pk:500 mL
- Description:Panserin PX40
Panserin PX40 is a serum-free complete medium for the cultivation of variety of cells.
- Ready to use
Based on RPMI 1640/DMEM/F-12, trace elements, albumin, cholesterol, lipids, vitamins and hormones were added to the medium. The medium does not contain any growth factors.