Serum-free media, Panserin 701
Catalog # PANBP04-710701
Supplier: PAN BIOTECH
CAS Number:
Specifications
- Media Format:Liquid
- Pk:500 mL
- Description:Panserin 701, with L-Glutamine and phytohemagglutinin (PHA)
- Seq:02
Specifications
About this item
Panserin 701 is a fully defined, complete ready-to-use serum-free medium for the cultivation of lymphocytes from whole blood.
- Store at –20 °C
Based on Iscove's MEM the medium is enriched with additional trace elements, albumin, cholesterol, lipids and vitamins. It contains the mitogen phytohemagglutinin (PHA) for a growth stimulation of lymphocytes.