Order Entry
Singapore
ContactUsLinkComponent
Dichloromethane ≥99.9% stabilised, B&J Brand™ for HPLC, for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™
Irritant
Health
Dichloromethane ≥99.9% stabilised, B&J Brand™ for HPLC, for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™
Catalog # BJAA300-4
Supplier:  BURDICK JACKSON
CAS Number:  
undefined
Irritant
Health
Dichloromethane ≥99.9% stabilised, B&J Brand™ for HPLC, for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™
Catalog # BJAA300-4
Supplier:  BURDICK JACKSON
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    16 L
  • Seq:
    5254

Specifications

Specification Test Results

  • Acidity (as HCl):
    1 mg/L max.
  • Chloride:
    10 mg/L max.
  • Electron Capture GC:
    10 ng/L max.
  • GC Analysis:
    99.9 % min.
  • Refractive Index (20ºC):
    1.4236 - 1.4246
  • Residue:
    1 mg/L max.
  • UV Cutoff:
    233 nm max.
  • Water by Karl Fischer Titration:
    0.01 % max.

Specification Test Results

About this item

 :
 :84.93 g/mol
Merck Index:12,06140
Boiling Pt:40 °C
Melting Pt:–95 °C
Density:1,33 g/cm³
Flash Pt: Does not flash
EINECS:200-838-9
UN:1593
ADR:6.1,III
 :Stabilised with cyclohexene
 :GHS07
 :GHS08
Storage Temperature:Ambient
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881