Order Entry
Singapore
ContactUsLinkComponent
Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution
Irritant
Environment
Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution
Catalog # RICC7000-16
Supplier:  Ricca Chemical
CAS Number:  
undefined
Irritant
Environment
Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution
Catalog # RICC7000-16
Supplier:  Ricca Chemical
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

About this item

 :
 :169.87 g/mol
Storage Temperature:Ambient
EINECS:231-853-9
UN:3082
ADR:9,III
 :GHS07
 :GHS09
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414